Order Entry
Netherlands
ContactUsLinkComponent
Disposable volume capillaries 20 µl, CE conformity-certified, borosilicate glass 3.3, ISO 7550, adjusted to sprue = In from end to end, conformity-certified, accuracy = ± 0.5 % 100 pieces per pack 1 * 100 items
Catalog # HECH40551018
New Product
Disposable volume capillaries 20 µl, CE conformity-certified, borosilicate glass 3.3, ISO 7550, adjusted to sprue = In from end to end, conformity-certified, accuracy = ± 0.5 % 100 pieces per pack 1 * 100 items
Catalog # HECH40551018