80 PPB OF I- CS MO IN NAOH 0.1M 1 * 100 mL
Catalog # CMPLRM110792L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
80 PPB OF I- CS MO IN NAOH 0.1M 1 * 100 mL
Catalog # CMPLRM110792L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
VWR® Q Path® Labofix Cytology Fixative
Supplier: VWR Chemicals