Order Entry
Netherlands
ContactUsLinkComponent
Y-connector, 10 pieces, 4.8 mm ID - 9.5 mm OD For tubing with 4.8 - 6.5 mm ID PVDF/ Kynar - FDA USP Class IV, FDA CFR 177.2510. 1 * 10 items
Catalog # WATS84-012-010
Supplier:  WATSON MARLOW
Y-connector, 10 pieces, 4.8 mm ID - 9.5 mm OD For tubing with 4.8 - 6.5 mm ID PVDF/ Kynar - FDA USP Class IV, FDA CFR 177.2510. 1 * 10 items
Catalog # WATS84-012-010
Supplier:  WATSON MARLOW
Frequently Bought Together