PRIME-XV Dendritic Cell Maturation CDM is a chemically-defined, animal component-free medium developed for the in vitro differentiation of monocytes into immature dendritic cells (iDCs) and subsequent maturation to produce mature dendritic cells (mDC 1 * 500 mL
Catalog # SHBT91146
Supplier: Shenandoah Biotechnology