Order Entry
Netherlands
ContactUsLinkComponent
,,11 components: 10mg/l each of Au Hf Ir Os Pd Pt Rh Ru Sb Sn Te in HNO3 5% HCl 5 ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM105807L5
,,11 components: 10mg/l each of Au Hf Ir Os Pd Pt Rh Ru Sb Sn Te in HNO3 5% HCl 5 ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM105807L5
Frequently Bought Together