2,7-Dibromocarbazole
Glass bottle, 5 g
467780050.
: 2,7-Dibromo-9H-carbazole
- :136630-39-2
- :C₁₂H₇Br₂N
- :MFCD09033507
- :325.00 g/mol
- :Ambient
- :2811
- :6.1,III
- Appearance:white to yellow to beige powder and or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:230°C to 234°C
- Pack type:Glass bottle
- Pk:5 g
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



