Order Entry
Netherlands
ContactUsLinkComponent
,,4 components: 500mg/l each of Chlorides (Cl-) Nitrates (NO3-) Phosphates (PO43-) Sulphates (SO42-) in Water ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 200 mL
Catalog # CMPLRM096013L2
,,4 components: 500mg/l each of Chlorides (Cl-) Nitrates (NO3-) Phosphates (PO43-) Sulphates (SO42-) in Water ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 200 mL
Catalog # CMPLRM096013L2
Frequently Bought Together