SARS-CoV-2 (COVID-19) Nucleocapsid Antibody [3863] 1 * 0,1 mg
Catalog # PRSI35-711
Supplier: ProSci Inc.
SARS-CoV-2 (COVID-19) Nucleocapsid Antibody [3863] 1 * 0,1 mg
Catalog # PRSI35-711
Supplier: ProSci Inc.
Frequently Bought Together
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection