SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N002] 1 * 0,1 mg
Catalog # PRSI10-536
Supplier: ProSci Inc.
SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N002] 1 * 0,1 mg
Catalog # PRSI10-536
Supplier: ProSci Inc.
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Petri Dish, PS
Supplier: VWR Collection
J.T.Baker® Plate, SRW Magnetic
Supplier: Avantor
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
96-Well cell culture plates, Falcon®
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: TENAK