Order Entry
Netherlands
ContactUsLinkComponent
7 components: 10mg/l each of Au Ir Os Pd Pt Rh Ru in HCl 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM091209L1
7 components: 10mg/l each of Au Ir Os Pd Pt Rh Ru in HCl 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM091209L1
Frequently Bought Together