NOTCH1 RECOMBINANT MONOCLONAL ANTIBODY (E6) (RABBIT IGGΛ) 1 * 200 µG
ENZOENZABS6480200
NOTCH1 RECOMBINANT MONOCLONAL ANTIBODY (E6) (RABBIT IGGΛ) 1 * 200 µG
ENZOENZABS6480200
Frequently Bought Together
Microtubes, with thread
: SARSTEDT
Tissue vial box, blue, WHEATON® CryoFILE
: DWK Life Sciences
VWR®, Ice Pans, with or without Lid
: VWR Collection
Cellulose chromatography papers, Whatman™
: Whatman products (Cytiva)
PCR tubes, 0.2 ml, with flat or domed cap, ABgene®
: Thermo Fisher Scientific
Racks and lids for storage 2D barcoded tubes, Matrix™
: THERMO MATRIX TECHNOLOGIES
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
: Bel-Art Products, a Part of SP
Micro packaging vials with thread, Nalgene®
: Thermo Fisher Scientific
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES
Storage Boxes, PP, with Lid
: KOEHLER TECHNISCHE PRODUKTEN
NANOPLATE 26K 240-WELL 1 * 10 items
: QIAGEN
AcroPrep™ 24-Well Receiver Plate
: Cytiva (Formerly Pall Lab)