Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C-MYC (HUMAN) RECOMBINANT MONOCLONAL ANTIBODY (9E10) (MOUSE IGG2BΚ) 1 * 200 µG
Catalog # ENZOENZABS4700200
Supplier:
ENZO LIFE SCIENCES
Expand image
C-MYC (HUMAN) RECOMBINANT MONOCLONAL ANTIBODY (9E10) (MOUSE IGG2BΚ) 1 * 200 µG
Catalog # ENZOENZABS4700200
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection