0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
PALBOCICLIB (FREE BASE) 1 * 2 g
ENZOENZCHM2322000
:
Enzo Life Sciences
PALBOCICLIB (FREE BASE) 1 * 2 g
ENZOENZCHM2322000
:
Enzo Life Sciences
Frequently Bought Together
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, chloride, Spectroquant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
pH indicator papers, MQuant®, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Reagents for copper determination using Lovibond® MD photometers
: Lovibond
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY