CERITINIB (FREE BASE) 1 * 100 mg
Catalogus # ENZOENZCHM2180100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Acetic acid 99% ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Test strips for glucose, MQuant®, Supelco®
Supplier: Merck
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Hydrochloric acid 1 mol/l (1 N) for biopharmaceutical production
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
VWR® Q Path® Labofix Cytology Fixative
Supplier: VWR Chemicals
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
Powder pillows, reagents for Hach instruments
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach