Bestelling invoeren
Netherlands
Orders LinkContactUsLinkComponent
Nocodazole ≥98%

Nocodazole ≥98%

50 mg
Catalog # ENZOBMLT1010050
Supplier:   Enzo Life Sciences
Nocodazole ≥98%
Catalog # ENZOBMLT1010050
Supplier:  Enzo Life Sciences
CAS Number:  31430-18-9
Nocodazole ≥98%
Nocodazole ≥98%

50 mg

CAS Number:  31430-18-9
Price
509,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Microtubule inhibitor

Synthetic reversible microtubule inhibitor.

Synonyms:  Methyl (6-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-yl)carbamate
  • CAS Number:31430-18-9
  • Formula:C₁₄H₁₁N₃O₃S
  • MDL Number:MFCD00005588
  • Molecular Weight:301.32 g/mol
Specifications

Specifications

  • VPE:
    50 mg

Specifications