Order Entry
Netherlands
Orders LinkContactUsLinkComponent
BML-111 ≥99%

BML-111 ≥99%

5 mg
Catalog # ENZOBMLRA1080005
Supplier:   Enzo Life Sciences
BML-111 ≥99%
Catalog # ENZOBMLRA1080005
Supplier:  Enzo Life Sciences
CAS Number:  78606-80-1
BML-111 ≥99%
BML-111 ≥99%

5 mg

CAS Number:  78606-80-1
Price
379,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).

Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).

Synonyms:  5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
  • CAS Number:78606-80-1
  • Formula:C₈H₁₆O₅
  • Molecular Weight:192.2 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Purity:
    ≥99%

Specifications

  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.