BML-111 ≥99%
1 mg
ENZOBMLRA1080001
Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).
Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).
: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
- :78606-80-1
- :C₈H₁₆O₅
- :192.2 g/mol
- :Ambient
- Purity:≥99%
- Pk:1 mg



