Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2-Methoxy-3-nitrobenzonitrile 98%

2-Methoxy-3-nitrobenzonitrile 98%

Bottle, 5 g
Catalog # APOSOR909773-5G
Supplier:   Apollo Scientific
2-Methoxy-3-nitrobenzonitrile 98%
Catalog # APOSOR909773-5G
Supplier:  Apollo Scientific
CAS Number:  121001-06-7
2-Methoxy-3-nitrobenzonitrile 98%

Bottle, 5 g

CAS Number:  121001-06-7
Price
652,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:121001-06-7
  • Formula:C₈H₆N₂O₃
  • MDL Number:MFCD18398370
  • Molecular Weight:178.145 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.