Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Rhod-FF AM

Plastic bottle, 1 mg
Catalogus # AATB21077
Irriterend
Leverancier:   AAT Bioquest
Rhod-FF AM
Catalogus # AATB21077
Leverancier:  AAT Bioquest
CAS-nummer:  
Rhod-FF AM
Rhod-FF AM

Plastic bottle, 1 mg

CAS-nummer:  
Price
469,00 €
Log in voor beschikbaarheid
1 * 1 mg
Product Details & Documents

Calcium measurement is critical for numerous biological investigations.

  • Calcium-dependent fluorescence
  • Low binding affinity
  • Cell permeant

 : For research use only.

  • Molecular Weight:1145.9 g/mol
  • ADR:5,II
Specificaties

Specificaties

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specificaties