Order Entry
Netherlands
ContactUsLinkComponent
Hazardous material work station with fresh air curtain, without working surface ventilation testing of the air equipment in accordance with EN 14175-3 (5.4.4) Exhaust and fresh air monitoring with visual and audible alarm, Interior lighting aranged a 1 * 1 items
Catalog # MSGA40572
Supplier:  asecos
Hazardous material work station with fresh air curtain, without working surface ventilation testing of the air equipment in accordance with EN 14175-3 (5.4.4) Exhaust and fresh air monitoring with visual and audible alarm, Interior lighting aranged a 1 * 1 items
Catalog # MSGA40572
Supplier:  asecos
Frequently Bought Together