Order Entry
Netherlands
ContactUsLinkComponent
Fractogel® EMD SO3? (M) strong cation exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm) 1x 500ml Parameter : 8 bar max. pressure Sterilität : sterile (Caustic Stable) Mittlere Partikelgröße : 40-90 µm 1 * 500 mL
Catalog # MILX1168820500
Supplier:  Merck
Fractogel® EMD SO3? (M) strong cation exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm) 1x 500ml Parameter : 8 bar max. pressure Sterilität : sterile (Caustic Stable) Mittlere Partikelgröße : 40-90 µm 1 * 500 mL
Catalog # MILX1168820500
Supplier:  Merck
Frequently Bought Together