Cerex® Atlas® SPE Columns
Product Details & Documents
A hydrophilic/Lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species, also comes in larger particle for more viscous samples. A direct alternative to Waters Oasis® HLB and Phenomenex Strata X.
- Mode: Reversed-phased
Atlas® a hydrophilic-lipophilic balanced polymeric sorbent is a N-vinylpyrrolidone-divinylbenzene co-polymer. It provides selectivity for both polar and non-polar analytes thus allowing the user to extract a variety of samples with ease. The divinylbenzene lipophilic back bone provides hydrophobic selectivity while the pyrrolidone hydrophilic chemistry will help increase interactions with polar functional groups of the analyte
Specifications
- Particle Size:60 µm
- Capacity:6 ml
- Description:Cerex Atlas® column
- Seq:09
- Bed weight:150 mg
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



