Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Cerex® SPE Columns HPWCX

Cerex® SPE Columns HPWCX

Column Cerex HPWCX, 25 µm, 1 ml, 10 mg
Catalog # TCAT30174772
Supplier:   Tecan
Cerex® SPE Columns HPWCX
Catalog # TCAT30174772
Supplier:  Tecan
Cerex® SPE Columns HPWCX

Column Cerex HPWCX, 25 µm, 1 ml, 10 mg

Price
Inquire For Price
Log in for availability
1 * 96 ST
Product Details & Documents

Polymer monodisperse weak cation exchange chemistry, used for retention of strong bases in biological fluids. Ideal for strong base extraction for drug monitoring, screening, identification, confirmation, and quantitation.

  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Specifications
  • Particle Size:
    25 µm
  • Capacity:
    1 ml
  • Description:
    Column Cerex HPWCX
  • Seq:
    01
  • Bed weight:
    10 mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.