TRYPSIN MS APPROVED F. PORK PANCREAS 1 * 100 µG
Catalog # SERA37286.01
Supplier: Serva
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Methyl acetate anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Ethylene glycol, high purity
Supplier: VWR Chemicals
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor