Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF124
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF124
Supplier: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Gas lighter, Varaflame
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Accessories for tube mill control
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Accessories for Thermo CO₂ resistant orbital shaker
Supplier: Thermo Fisher Scientific
Magnetic stirring bars
Supplier: VELP SCIENTIFIC
VWR®, Rotating mixer XL
Supplier: VWR Collection
Accessories for hotplates, VHP-C Series
Supplier: IKA
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Magnetic hotplate stirrers, C-MAG HS 7 control
Supplier: IKA
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Multi-position magnetic stirrers, MULTISTIRRER series
Supplier: VELP SCIENTIFIC
Overhead stirrer tools, dissolver stirrers
Supplier: IKA
Accessories for analytical Mill, A 11 basic
Supplier: IKA
Stirrers, propeller, with three paddles
Supplier: BOCHEM