Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CF4M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CF4M
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
VWR® VWB2, Water Baths
Supplier: VWR Collection
Accessories for vortex mixers, MS 3
Supplier: IKA
Globe stirrer couplings, BOLA
Supplier: Bohlender
VWR®, Tube rollers
Supplier: VWR Collection
Sieves for POLYMIX® system PX-MFC 90 D
Supplier: KINEMATICA
MaxQ™ 8000 Incubating Shakers
Supplier: Thermo Fisher Scientific
DISPERSING TOOL S 25 EC-T-C-25G-ST 1 * 1 items
Supplier: IKA
Grinding jar EasyFit for Planetary Ball Mills, aluminium oxide, 250ml 1 * 1 items
Supplier: Retsch GmbH
ORBITAL SHAKING PLATFORM 1 * 1 items
Supplier: Cole Parmer
Grinding balls for XRD-mill McCrone
Supplier: Retsch GmbH
VWR®, Accessories for Microplate Shakers and Orbital/ Mini Shakers
Supplier: VWR Collection
Overhead stirrer tools, propeller, centrifugal and paddle-type impellers
Supplier: Heidolph Instruments GmbH & Co.KG
Racks for orbital shakers SM 30 A, SM 30 A control, SM 30 B and SM 30 B control
Supplier: Edmund Bühler GmbH
Pierceable septums, pack of 10 pcs 1 * 10 items
Supplier: VELP SCIENTIFIC
Test tubes, glass, with screw cap
Supplier: witeg Labortechnik