Order Entry
Netherlands
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
  SARBMCA116S3S0034H
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
  SARBMCA116S3S0034H
Frequently Bought Together