potassium (1-tert-butoxycarbonyl-2,3,6,7-tetrahydroazepin-4-yl)-trifluoro-boranuide 1 * 1 g
Catalog # PHBLPBZT152-8-1G
Supplier: Pharmablock (USA)
potassium (1-tert-butoxycarbonyl-2,3,6,7-tetrahydroazepin-4-yl)-trifluoro-boranuide 1 * 1 g
Catalog # PHBLPBZT152-8-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Pentanol ≥98.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, Technical Grade
Supplier: Thermo Fisher Scientific