methyl cis-3-(methylamino)cyclobutanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBZ4520-1-1G
Supplier: Pharmablock (USA)
methyl cis-3-(methylamino)cyclobutanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBZ4520-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol absolute 99.8% for analysis
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
n-Heptane 99+% for HPLC
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals