O1-tert-butyl O3-methyl (3S,5R)-5-aminopiperidine-1,3-dicarboxylate hydrochloride 1 * 1 g
PHBLPBZ1810-1-1G
O1-tert-butyl O3-methyl (3S,5R)-5-aminopiperidine-1,3-dicarboxylate hydrochloride 1 * 1 g
PHBLPBZ1810-1-1G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals