(2,5-DIOXOPYRROLIDIN-1-YL) 3-[2-[2-[2-[2 1 * 1 g
Catalog # PHBLPBY2035-1G
Supplier: Pharmablock (USA)
(2,5-DIOXOPYRROLIDIN-1-YL) 3-[2-[2-[2-[2 1 * 1 g
Catalog # PHBLPBY2035-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Tolueen ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Xyleen ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine USP, Ph. Eur., BP
Supplier: VWR Chemicals