N-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethyl]-2-[2-(2,6-dioxo-3-piperidyl)-1,3-dioxo-isoindolin-4-yl]oxy-acetamide hydrochloride 1 * 1 g
Catalog # PHBLPBY1810-1-1G
Supplier: Pharmablock (USA)
N-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethyl]-2-[2-(2,6-dioxo-3-piperidyl)-1,3-dioxo-isoindolin-4-yl]oxy-acetamide hydrochloride 1 * 1 g
Catalog # PHBLPBY1810-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals