N-[2-[2-(2-hydroxyethoxy)ethoxy]ethyl]-5-(2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl)pentanamide 1 * 1 g
Catalog # PHBLPBY1546-1G
Supplier: Pharmablock (USA)
N-[2-[2-(2-hydroxyethoxy)ethoxy]ethyl]-5-(2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl)pentanamide 1 * 1 g
Catalog # PHBLPBY1546-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific