2-[2-[2-[2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1489-1G
Supplier: Pharmablock (USA)
2-[2-[2-[2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1489-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Diethyl Ether stabilized with &sim 6 ppm of BHT (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 mL
Supplier: PanReac AppliChem
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals