2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethylamino]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1466-1G
Supplier: Pharmablock (USA)
2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethylamino]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1466-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% (by GC) stabilised, Puriss., meets analytical specification of Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide 99%, Reagent Grade
Supplier: Honeywell Chemicals
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck