2-[2-[2-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1317-1G
Supplier: Pharmablock (USA)
2-[2-[2-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1317-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
o-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetylacetone, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.9%
Supplier: Honeywell Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific