2-[2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1070-1G
Supplier: Pharmablock (USA)
2-[2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY1070-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals