5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-(2-azidoethoxy)ethoxy]ethyl]pentanamide 1 * 1 g
Catalog # PHBLPBY0722-1G
Supplier: Pharmablock (USA)
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-(2-azidoethoxy)ethoxy]ethyl]pentanamide 1 * 1 g
Catalog # PHBLPBY0722-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine 98.0-101.0% (by alkalimetry) ACS, Reag. Ph. Eur.
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Pyridine ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 40…60 °C ACS
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
Dichloromethane
Supplier: Roth Carl