6-(2,5-dioxopyrrol-1-yl)-N-[(1S)-1-[[(1S)-2-[4-(hydroxymethyl)anilino]-1-methyl-2-oxo-ethyl]carbamoyl]-2-methyl-propyl]hexanamide 1 * 1 g
Catalog # PHBLPBY0633-1G
Supplier: Pharmablock (USA)
6-(2,5-dioxopyrrol-1-yl)-N-[(1S)-1-[[(1S)-2-[4-(hydroxymethyl)anilino]-1-methyl-2-oxo-ethyl]carbamoyl]-2-methyl-propyl]hexanamide 1 * 1 g
Catalog # PHBLPBY0633-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals