2-[2-[2-[2-[2-(2-tetrahydropyran-2-yloxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY0598-1G
Supplier: Pharmablock (USA)
2-[2-[2-[2-[2-(2-tetrahydropyran-2-yloxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
Catalog # PHBLPBY0598-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol absolute ≥99%
Supplier: KLINIPATH
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Supplier: Avantor
Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…65 °C, Technical Grade
Supplier: Thermo Fisher Scientific