9H-fluoren-9-ylmethyl N-[(1S)-1-[[(1S)-2-[4-(hydroxymethyl)anilino]-1-methyl-2-oxo-ethyl]carbamoyl]-2-methyl-propyl]carbamate 1 * 1 g
Catalog # PHBLPBY0536-1G
Supplier: Pharmablock (USA)
9H-fluoren-9-ylmethyl N-[(1S)-1-[[(1S)-2-[4-(hydroxymethyl)anilino]-1-methyl-2-oxo-ethyl]carbamoyl]-2-methyl-propyl]carbamate 1 * 1 g
Catalog # PHBLPBY0536-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Toluene ≥99.7% (by GC), CHROMASOLV™ for pesticide residue analysis acc. to FDA, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N-Bromosuccinimide 99%
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific
Methanol 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.3%, Laboratory Reagent
Supplier: Honeywell Chemicals
Formaldehyde 37% (w/w) in aqueous solution stabilised
Supplier: Thermo Fisher Scientific
1-Butanol for HPLC
Supplier: Thermo Fisher Scientific