2-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]ethanol 1 * 1 g
PHBLPBY0372-1G
Frequently Bought Together
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
1,2-Dichloroethane 99+% for spectroscopy
: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Diethyl ether, anhydrous ≥99% stabilised ACS
: Thermo Fisher Scientific
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
: VWR Chemicals
Liquid for manometer with sloping tube
: VWR Chemicals
Ethyl acetate 99.5% for analysis
: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals