methyl (2S,3S)-2-methyl-1-[(1S)-1-phenylethyl]pyrrolidine-3-carboxylate 1 * 1 g
Catalog # PHBLPBXAA1161-1G
Supplier: Pharmablock (USA)
methyl (2S,3S)-2-methyl-1-[(1S)-1-phenylethyl]pyrrolidine-3-carboxylate 1 * 1 g
Catalog # PHBLPBXAA1161-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
Acetone for residue analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Petroleum spirit, 50…70 °C ≥90%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
Acetone 99.6% ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Fisher Scientific