tert-butyl cis-3-hydroxy-4-(methylamino)pyrrolidine-1-carboxylate 1 * 1 g
PHBLPBXA3182-1G
tert-butyl cis-3-hydroxy-4-(methylamino)pyrrolidine-1-carboxylate 1 * 1 g
PHBLPBXA3182-1G
Frequently Bought Together
n-Hexane
: Roth Carl
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
1,5-Diaminopentane 98%
: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
: Thermo Fisher Scientific
Triethanolamine (trolamine) BP, Ph. Eur., USP-NF, pharma grade
: PanReac AppliChem
2-Methylbutane 99+%, extra pure
: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals
2-Ethoxyethanol 99%, extra pure
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Acetone ≥99.5%, Laboratory Reagent
: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
: Thermo Fisher Scientific