O1-tert-butyl O2-methyl 5-(3-hydroxypropyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBXA300337-1G
Leverancier: Pharmablock (USA)
O1-tert-butyl O2-methyl 5-(3-hydroxypropyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBXA300337-1G
Leverancier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Leverancier: Merck
Potassium bromide, Uvasol® for IR spectrophotometry, Supelco®
Leverancier: Merck
Water USP, JP, Ph. Eur., purified
Leverancier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Leverancier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Leverancier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Leverancier: Merck
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Leverancier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Leverancier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Methanol, EMPLURA®, Supelco®
Leverancier: Merck
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals