rel-(3aR,5r,6aS)-5-methyl-2,3,3a,4,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-5-ol hydrochloride 1 * 1 g
Catalog # PHBLPBX1028-1-1G
Supplier: Pharmablock (USA)
rel-(3aR,5r,6aS)-5-methyl-2,3,3a,4,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-5-ol hydrochloride 1 * 1 g
Catalog # PHBLPBX1028-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
1-Chloro-2,4-dinitrobenzene 99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Chemicals
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
1-Butanol NF
Supplier: Spectrum Chemical
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific