2-[N-(2-hydroxyethyl)-3-methoxy-4-nitroso-anilino]ethanol 1 * 1 g
Catalog # PHBLPBWZ038-1G
Supplier: Pharmablock (USA)
2-[N-(2-hydroxyethyl)-3-methoxy-4-nitroso-anilino]ethanol 1 * 1 g
Catalog # PHBLPBWZ038-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific