4,4,5,5-tetramethyl-2-(3-oxabicyclo[4.1.0]heptan-6-yl)-1,3,2-dioxaborolane 1 * 1 g
Catalog # PHBLPBWBD0155-1G
Supplier: Pharmablock (USA)
4,4,5,5-tetramethyl-2-(3-oxabicyclo[4.1.0]heptan-6-yl)-1,3,2-dioxaborolane 1 * 1 g
Catalog # PHBLPBWBD0155-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Pyridine ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific