tert-butyl 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBWB100432-1G
Supplier: Pharmablock (USA)
tert-butyl 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBWB100432-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2-Dichloroethane ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
n-Hexane
Supplier: Roth Carl
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific