O1-tert-butyl O3-ethyl (3S,4R)-4-hydroxypiperidine-1,3-dicarboxylate 1 * 1 g
Catalog # PHBLPBWB100416-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O3-ethyl (3S,4R)-4-hydroxypiperidine-1,3-dicarboxylate 1 * 1 g
Catalog # PHBLPBWB100416-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals